CAS RN 150058-27-8
IUPAC Navn:
methyl 2-ethoxy-1H-1,3-benzodiazole-4-carboxylate
Synonymer:
methyl 2-ethoxy-1H-1,3-benzodiazole-4-carboxylate
Molekylvægt (g/mol):
220.23
Molekylformel:
C11H12N2O3
InChi Key:
MOPLKVMMSFGZIR-UHFFFAOYSA-N
SMILES:
CCOC1=NC2=C(C=CC=C2N1)C(=O)OC
1
–
2
af
2
Resultater
1
2-Ethoxy-3H-benzimidazole-4-carboxylic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Ethoxy-3H-benzimidazole-4-carboxylic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.