CAS RN 149402-51-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
HPPH, MedChemExpress
MedChemExpress HPPH (Photochlor) is a second generation photosensitizer, which acts as a photodynamic therapy (PDT) agent.