CAS RN 1482-70-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
(5Alpha)-Androstane-3,11,17-trione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.