CAS RN 14807-96-6
IUPAC Navn:
trimagnesium(2+) bis(oxosilanebis(olate)) dihydroxysilanoylolate
Synonymer:
TALC talc: hydrated magnesium silicate
Molekylvægt (g/mol):
379.26
Molekylformel:
H2Mg3O12Si4
InChi Key:
KNXVOGGZOFOROK-UHFFFAOYSA-N
SMILES:
[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O
1
–
1
af
1
Resultater
1
Talc, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.