CAS RN 148-72-1
IUPAC Navn:
(3S)-3-ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]oxolan-2-one; nitric acid
Synonymer:
(3S)-3-ethyl-4-[(3-methylimidazol-4-yl)methyl]oxolan-2-one; nitric acid
Molekylvægt (g/mol):
271.27
Molekylformel:
C11H17N3O5
InChi Key:
PRZXEPJJHQYOGF-LQRGNCEWSA-N
SMILES:
O[N+]([O-])=O.CC[C@H]1C(CC2=CN=CN2C)COC1=O
1
–
3
af
3
Resultater
1
Pilocarpine Nitrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Pilocarpine Nitrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Pilocarpine Nitrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.