missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Pilocarpine Nitrate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Pilocarpine Nitrate |
| CAS | 148-72-1 |
| Molekylær formel | C11 H16 N2 O2 . H N O3 |
| Mængde | 5 g |
| Synonym | 2(3H)-Furanone','3-ethyldihydro-4-[(1-methyl-1H-imidazol-5-yl)methyl]-','(3S,4R)-','nitrate (1:1), 2(3H)-Furanone','3-ethyldihydro-4-[(1-methyl-1H-imidazol-5-yl)methyl]-','(3S,4R)-','mononitrate (9CI), 2(3H)-Furanone','3-ethyldihydro-4-[(1-methyl-1H-imidazol-5-yl)methyl]-','(3S-cis)-','mononitrate, Pilocarpine','mononitrate (8CI), (+)-Pilocarpine nitrate, Pilagan, Pilocarpine nitrate, Pilocarpine subnitrate |
| InChI formel | InChI=1S/C11H16N2O2.HNO3/c1-3-10-8(6-15-11(10)14)4-9-5-12-7-13(9)2;2-1(3)4/h5,7-8,10H,3-4,6H2,1-2H3;(H,2,3,4)/t8-,10-;/m0./s1 |
| SMIL | CC[C@H]1[C@@H](Cc2cncn2C)COC1=O.O[N+](=O)[O-] |
| IUPAC navn | (3S,4R)-3-ethyl-4-[(3-methylimidazol-4-yl)methyl]oxolan-2-one;nitric acid |
| Molekylvægt (g/mol) | 271.27 |
| Formel vægt | 271.1168 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?