CAS RN 1478663-96-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(4S)-3-[(2R)-2-[(R)-[4-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]phenyl][(4-fluorophenyl)amino]methyl]-4-[2-(4-fluorophenyl)-5,5-dimethyl-1,3-dioxan-2-yl]-1-oxobutyl]-4-phenyl-2-oxazolidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(4S)-3-[(2R)-2-[(R)-[4-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]phenyl][(4-fluorophenyl)amino]methyl]-4-[2-(4-fluorophenyl)-5,5-dimethyl-1,3-dioxan-2-yl]-1-oxobutyl]-4-phenyl-2-oxazolidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(4S)-3-[(2R)-2-[(R)-[4-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]phenyl][(4-fluorophenyl)amino]methyl]-4-[2-(4-fluorophenyl)-5,5-dimethyl-1,3-dioxan-2-yl]-1-oxobutyl]-4-phenyl-2-oxazolidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.