CAS RN 147698-05-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(S)-1-tert-Butyl 6-methyl 2-((tert-butoxycarbonyl)amino)hexanedioate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(S)-1-tert-Butyl 6-methyl 2-((tert-butoxycarbonyl)amino)hexanedioate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(S)-1-tert-Butyl 6-methyl 2-((tert-butoxycarbonyl)amino)hexanedioate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.