missing translation for 'onlineSavingsMsg'
Få mere at vide

(S)-1-tert-Butyl 6-methyl 2-((tert-butoxycarbonyl)amino)hexanedioate, TRC

Artikelnummer. 30347793
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30347793 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347793 Supplier Toronto Research Chemicals Supplier No. TRCB117825250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (S)-1-tert-Butyl 6-methyl 2-((tert-butoxycarbonyl)amino)hexanedioate
CAS 147698-05-3
Molekylær formel C12 H4 Cl6 O
Mængde 250 mg
Synonym 2S)-[[(1,1-dimethylethoxy)carbonyl]amino]-hexanedioic Acid 1-(1,1-Dimethylethyl) 6-Methyl Ester
InChI formel InChI=1S/C16H29NO6/c1-15(2,3)22-13(19)11(9-8-10-12(18)21-7)17-14(20)23-16(4,5)6/h11H,8-10H2,1-7H3,(H,17,20)/t11-/m0/s1
SMIL [C@@H](NC(OC(C)(C)C)=O)(C(OC(C)(C)C)=O)CCCC(OC)=O
IUPAC navn 2,3,6-trichloro-4-(2,3,4-trichlorophenyl)phenol
Molekylvægt (g/mol) 331.41
Formel vægt 331.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.