CAS RN 1459721-06-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
rel-(2R,3aR,5S,6S,6aS)-4-Methylhexahydro-2H-2,5-methanofuro[3,2-b]pyrrol-6-yl 2-hydroxy-2,2-di(thiophen-2-yl)acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.