CAS RN 1453807-04-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(5Z,8Z,11Z,14Z)-5,8,11,14-Eicosatetraenoic Acid 2-[[(1,1-Dimethylethoxy)carbonyl]amino]ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(5Z,8Z,11Z,14Z)-5,8,11,14-Eicosatetraenoic Acid 2-[[(1,1-Dimethylethoxy)carbonyl]amino]ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(5Z,8Z,11Z,14Z)-5,8,11,14-Eicosatetraenoic Acid 2-[[(1,1-Dimethylethoxy)carbonyl]amino]ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.