CAS RN 14516-46-2
IUPAC Navn:
palladium(2+) dipotassium tetrakis(iminomethanide)
Synonymer:
palladium(2+) dipotassium tetrakis(cyanide)
Molekylvægt (g/mol):
288.69
Molekylformel:
C4K2N4Pd
InChi Key:
IYDMNMSJMUMQBP-UHFFFAOYSA-N
SMILES:
[K+].[K+].[Pd++].[C-]#N.[C-]#N.[C-]#N.[C-]#N