CAS RN 144875-92-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
2-[(Acetyloxy)methyl]-Alpha,Alpha-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-[(Acetyloxy)methyl]-Alpha,Alpha-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-[(Acetyloxy)methyl]-Alpha,Alpha-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.