CAS RN 1446322-66-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Tocris Bioscience™ TAT-cyclo-CLLFVY
Selective HIF-1 dimerization inhibitor
2