CAS RN 1445385-02-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Lumicitabine, MedChemExpress
MedChemExpress Lumicitabine (ALS-008176) is an inhibitor of the respiratory syncytial virus (RSV) polymerase.