CAS RN 1436861-97-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Zidebactam, MedChemExpress
MedChemExpress Zidebactam (WCK-5107) is a potent β-lactamase inhibitor. Zidebactam also is a penicillin-binding protein2 (PBP2) inhibitor with an IC50 of 0.26 μg/mL.