Få mere at vide
Zidebactam, MedChemExpress
Artikelnummer. 16437717
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16437717 5 mg
16414988 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16437717 Leverandør Medchem Express Leverandørnr. HY120859
MedChemExpress Zidebactam (WCK-5107) is a potent β-lactamase inhibitor. Zidebactam also is a penicillin-binding protein2 (PBP2) inhibitor with an IC50 of 0.26 μg/mL.
Zidebactam (WCK-5107) is a potent β-lactamase inhibitor. Zidebactam also is a penicillin-binding protein2 (PBP2) inhibitor with an IC50 of 0.26 μg/mL.
Tekniske data
Kemisk navn eller materiale Zidebactam
CAS 1436861-97-0
Farve White
Renhedsgrad noter Research
Molekylær formel C13H21N5O7S
Mængde 5 mg
Synonym WCK-5107
Opløselighedsinformation DMSO : 250 mg/mL (638.73 mM; Need ultrasonic) ∣H2O : 50 mg/mL (127.75 mM; Need ultrasonic)
SMIL O=C([C@@H]1CCCNC1)NNC([C@@H]2CC[C@@H]3C[N@]2C(N3OS(=O)(O)=O)=O)=O
Molekylvægt (g/mol) 391.4
Formel vægt 391.4
Procent renhed 94.42%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation