CAS RN 143612-79-7
IUPAC Navn:
3-(4-chlorobutyl)-1H-indole-5-carbonitrile
Synonymer:
3-(4-chlorobutyl)-1H-indole-5-carbonitrile
Molekylvægt (g/mol):
232.71
Molekylformel:
C13H13ClN2
InChi Key:
NJJWMEJWFYRORL-UHFFFAOYSA-N
SMILES:
ClCCCCC1=CNC2=CC=C(C=C12)C#N
1
–
2
af
2
Resultater
1
3-(4-Chlorobutyl)indole-5-carbonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-(4-Chlorobutyl)indole-5-carbonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.