CAS RN 1431697-96-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
CCT241533 hydrochloride, MedChemExpress
MedChemExpress CCT241533 hydrochloride is a potent and selective CHK2 inhibitor with an IC50 of 3 nM and a Ki of 1.16 nM.