CAS RN 1430202-69-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ 2R,6R-Hydroxynorketamine hydrochloride
Enhances AMPA currents; decreases D-serine (a NMDA co-agonist); lacks ketamine-related side effects