CAS RN 143-62-4
IUPAC Navn:
4-[(1R,3aS,3bR,5aR,7S,9aS,9bS,11aR)-3a,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]-2,5-dihydrofuran-2-one
Synonymer:
digitoxigenin digitoxigenine echujetin uzarigenin thevetigenin cerberigenin evonogenin
Molekylvægt (g/mol):
374.52
Molekylformel:
C23H34O4
InChi Key:
XZTUSOXSLKTKJQ-CESUGQOBSA-N
SMILES:
C[C@]12CC[C@H]3[C@@H](CC[C@@H]4C[C@@H](O)CC[C@]34C)[C@@]1(O)CC[C@@H]2C1=CC(=O)OC1
1
–
3
af
3
Resultater
1
Digitoxigenin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Digitoxigenin, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Digitoxigenin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.