CAS RN 1428935-60-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
MedChemExpress Durvalumab
MedChemExpress Durvalumab (MEDI 4736) is an humanized anti-PD-L1 monoclonal antibody. Durvalumab (MEDI4736) completely blocks the binding of PD-L1 to both PD-1 and CD80, with IC50s of 0.1 and 0.04 nM, respectively.
2
MedChemExpress Durvalumab
MedChemExpress Durvalumab (MEDI 4736) is an humanized anti-PD-L1 monoclonal antibody. Durvalumab (MEDI4736) completely blocks the binding of PD-L1 to both PD-1 and CD80, with IC50s of 0.1 and 0.04 nM, respectively.