CAS RN 14166-21-3
IUPAC Navn:
octahydro-2-benzofuran-1,3-dione
Synonymer:
hexahydrophthalic anhydride HHPA
Molekylvægt (g/mol):
154.17
Molekylformel:
C8H10O3
InChi Key:
MUTGBJKUEZFXGO-UHFFFAOYNA-N
SMILES:
O=C1OC(=O)C2CCCCC12
1
–
1
af
1
Resultater
1
Trans-1,2-cyclohexanedicarboxylic Anhydride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.