CAS RN 1416561-90-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Tocris Bioscience™ O-Propargyl-puromycin
Puromycin analog for labeling newly synthesized proteins; is incorporated into nascent polypeptides
2
O-Propargyl-Puromycin, MedChemExpress
MedChemExpress O-Propargyl-Puromycin, an alkyne analog of puromycin, is a potent protein synthesis inhibitor.