CAS RN 1415564-68-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
ST-836 hydrochloride, MedChemExpress
MedChemExpress ST-836 hydrochloride (compound 34) is a potent dopamine receptor ligand with Ki values of 4.5 nM, 132 nM for D3 and D2, respectively. ST-836 hydrochloride has the potential for Parkinson’s disease.