CAS RN 1404456-53-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
GSK 2830371, MedChemExpress
MedChemExpress GSK 2830371 is a highly selective Wip1 phosphatase inhibitor with IC50 of 6 nM.