CAS RN 1402819-05-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(6-Bromo-2-ethyl-3-benzofuranyl)(3,5-dibromo-4-hydroxyphenyl)methanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(6-Bromo-2-ethyl-3-benzofuranyl)(3,5-dibromo-4-hydroxyphenyl)methanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.