CAS RN 1391053-23-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
6-Benzyloxy-1-cyclopropyl-1,4-dihydro-7-fluoro-4-oxo-3-quinolinecarboxylic Acid Benzyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6-Benzyloxy-1-cyclopropyl-1,4-dihydro-7-fluoro-4-oxo-3-quinolinecarboxylic Acid Benzyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.