CAS RN 1390658-68-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
N-Methyl-N-[3-[[[(8Beta)-6-(2-propen-1-yl)ergolin-8-yl]carbonyl]amino]propyl]carbamic Acid 1,1-Dimethylethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Methyl-N-[3-[[[(8b)-6-(2-propen-1-yl)ergolin-8-yl]carbonyl]amino]propyl]carbamic Acid 1,1-Dimethylethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Methyl-N-[3-[[[(8Beta)-6-(2-propen-1-yl)ergolin-8-yl]carbonyl]amino]propyl]carbamic Acid 1,1-Dimethylethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.