CAS RN 1386979-55-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Mirk-IN-1, MedChemExpress
MedChemExpress Mirk-IN-1 is a potent inhibitor of Dyrk1B(Mirk kianse) and Dyrk1A with IC50 of 68±48 nM and 22±8 nM respectively.