CAS RN 1380575-43-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Ceritinib dihydrochloride, MedChemExpress
MedChemExpress Ceritinib dihydrochloride (LDK378 dihydrochloride) is a selective, orally bioavailable and ATP-competitive ALK tyrosine kinase inhibitor with an IC50 of 200 pM. Ceritinib dihydrochloride (LDK378 dihydrochloride) also inhibits IGF-1R, InsR, and STK22D with IC50 values of 8, 7, and 23 nM, respectively. Ceritinib dihydrochloride (LDK378 dihydrochloride) shows great antitumor potency.