missing translation for 'onlineSavingsMsg'
Få mere at vide

Ceritinib dihydrochloride, MedChemExpress

Artikelnummer. 16452807
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16452807 50 mg -
16449277 5 mg -
16446866 100 mg -
16436728 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16452807 Leverandør Medchem Express Leverandørnr. HY15656A

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Ceritinib dihydrochloride (LDK378 dihydrochloride) is a selective, orally bioavailable and ATP-competitive ALK tyrosine kinase inhibitor with an IC50 of 200 pM. Ceritinib dihydrochloride (LDK378 dihydrochloride) also inhibits IGF-1R, InsR, and STK22D with IC50 values of 8, 7, and 23 nM, respectively. Ceritinib dihydrochloride (LDK378 dihydrochloride) shows great antitumor potency.

Ceritinib dihydrochloride (LDK378 dihydrochloride) is a selective, orally bioavailable and ATP-competitive ALK tyrosine kinase inhibitor with an IC50 of 200 pM. Ceritinib dihydrochloride (LDK378 dihydrochloride) also inhibits IGF-1R, InsR, and STK22D with IC50 values of 8, 7, and 23 nM, respectively. Ceritinib dihydrochloride (LDK378 dihydrochloride) shows great antitumor potency.

Tekniske data

Kemisk navn eller materiale Ceritinib dihydrochloride
CAS 1380575-43-8
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C28H38Cl3N5O3S
Mængde 50 mg
Synonym LDK378 dihydrochloride
Opløselighedsinformation DMSO : 100 mg/mL (158.46 mM; Need ultrasonic) ∣H2O : 10 mg/mL (15.85 mM; Need ultrasonic)
SMIL CC(C)OC1=CC(C2CCNCC2)=C(C)C=C1NC3=NC=C(Cl)C(NC4=CC=CC=C4S(=O)(C(C)C)=O)=N3.[H]Cl.[H]Cl
Molekylvægt (g/mol) 631.06
Formel vægt 631.06
Procent renhed 99.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.