CAS RN 1380316-03-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
EPZ004777 hydrochloride, MedChemExpress
MedChemExpress EPZ004777 hydrochloride is a potent, selective DOT1L inhibitor with an IC50 of 0.4 nM.