CAS RN 13795-24-9
IUPAC Navn:
benzyl 4-nitrophenyl carbonate
Synonymer:
benzyl 4-nitrophenyl carbonate
Molekylvægt (g/mol):
273.24
Molekylformel:
C14H11NO5
InChi Key:
QIXRWIVDBZJDGD-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(OC(=O)OCC2=CC=CC=C2)C=C1
1
–
2
af
2
Resultater
1
Benzyl 4-Nitrophenyl Carbonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Benzyl 4-Nitrophenyl Carbonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.