CAS RN 1365986-44-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Protein kinase inhibitors 1, MedChemExpress
MedChemExpress Protein kinase inhibitors 1 is a novel inhibitor of HIPK2 with an IC50 of 74 nM and Kd of 9.5 nM.