CAS RN 1359873-45-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
6-Ethyl-4-[4-(1,3,4-thiadiazol-2-yl)-1-piperazinyl]thieno[2,3-d]pyrimidine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6-Ethyl-4-[4-(1,3,4-thiadiazol-2-yl)-1-piperazinyl]thieno[2,3-d]pyrimidine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
6-Ethyl-4-[4-(1,3,4-thiadiazol-2-yl)-1-piperazinyl]thieno[2,3-d]pyrimidine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.