CAS RN 1357158-81-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
AKT-IN-1, MedChemExpress
MedChemExpress AKT-IN-1 is an allosteric AKT inhibitor with an IC50 of 1.042 μM.