CAS RN 1346604-65-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
4'-Ethyl-7',8'-dihydro-4'-hydroxy-spiro[1,3-dioxolane-2,6'(3'H)-[1H]pyrano[3,4-f]indolizine]-3',10'(4'H)-dione-d5, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4'-Ethyl-7',8'-dihydro-4'-hydroxy-spiro[1,3-dioxolane-2,6'(3'H)-[1H]pyrano[3,4-f]indolizine]-3',10'(4'H)-dione-d5, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.