Få mere at vide
4'-Ethyl-7',8'-dihydro-4'-hydroxy-spiro[1,3-dioxolane-2,6'(3'H)-[1H]pyrano[3,4-f]indolizine]-3',10'(4'H)-dione-d5, TRC
Artikelnummer. 30371790
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30371790 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371790 Leverandør Toronto Research Chemicals Leverandørnr. TRCE91731225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4'-Ethyl-7',8'-dihydro-4'-hydroxy-spiro[1,3-dioxolane-2,6'(3'H)-[1H]pyrano[3,4-f]indolizine]-3',10'(4'H)-dione-d5
CAS 1346604-65-6
Molekylær formel C15H12D5NO6
Mængde 25 mg
Synonym (±)-4'-Ethyl-7',8'-dihydro-4'-hydroxy-spiro[1,3-dioxolane-2,6'(3'H)-[1H]pyrano[3,4-f]indolizine]-3',10'(4'H)-dione-d5
InChI formel InChI=1S/C15H17NO6/c1-2-14(19)10-7-11-15(21-5-6-22-15)3-4-16(11)12(17)9(10)8-20-13(14)18/h7,19H,2-6,8H2,1H3/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C1(O)C(=O)OCC2=C1C=C3N(CCC34OCCO4)C2=O
Molekylvægt (g/mol) 312.33