CAS RN 1346602-61-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Succinyl Bis[(phenylimino)-2,1-ethanediyl]bis(3-maleimidopropanamide), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Succinyl Bis[(phenylimino)-2,1-ethanediyl]bis(3-maleimidopropanamide), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Succinyl Bis[(phenylimino)-2,1-ethanediyl]bis(3-maleimidopropanamide), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.