CAS RN 1341224-83-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
PF-4136309, MedChemExpress
MedChemExpress PF-4136309 is a potent, selective, and orally bioavailable CCR2 antagonist, with IC50s of 5.2 nM, 17 nM and 13 nM for human, mouse and rat CCR2.