CAS RN 1331908-61-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
L-Citrulline-d6, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
L-Citrulline-d6, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.