CAS RN 1330180-09-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
4-[4-[(5S)-5-Phthalimidomethyl-2-oxo-3-oxazolidinyl]phenyl]-3-morpholinone-d4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-[4-[(5S)-5-Phthalimidomethyl-2-oxo-3-oxazolidinyl]phenyl]-3-morpholinone-d4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.