Få mere at vide
4-[4-[(5S)-5-Phthalimidomethyl-2-oxo-3-oxazolidinyl]phenyl]-3-morpholinone-d4, TRC
Artikelnummer. 30410818
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30410818 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410818 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3858522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-[4-[(5S)-5-Phthalimidomethyl-2-oxo-3-oxazolidinyl]phenyl]-3-morpholinone-d4
CAS 1330180-09-0
Molekylær formel C22H15D4N3O6
Mængde 2.5 mg
Synonym 2-[[(5S)-2-Oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]-1H-isoindole-1,3(2H)-dione-d4
InChI formel InChI=1S/C22H19N3O6/c26-19-13-30-10-9-23(19)14-5-7-15(8-6-14)24-11-16(31-22(24)29)12-25-20(27)17-3-1-2-4-18(17)21(25)28/h1-8,16H,9-13H2/t16-/m1/s1/i5D,6D,7D,8D
SMIL [2H]c1c([2H])c(N2C[C@H](CN3C(=O)c4ccccc4C3=O)OC2=O)c([2H])c([2H])c1N5CCOCC5=O
IUPAC navn 2-[[(5S)-2-oxo-3-[2,3,5,6-tetradeuterio-4-(3-oxomorpholin-4-yl)phenyl]-1,3-oxazolidin-5-yl]methyl]isoindole-1,3-dione
Molekylvægt (g/mol) 425.43
Formel vægt 425.1525