CAS RN 13270-56-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
(S)-6-Methylnicotine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(S)-6-Methylnicotine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(S)-6-Methylnicotine (1 mg/mL in Methanol), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
(S)-6-Methylnicotine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.