CAS RN 132172-61-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
1,2-Dioleoyl-3-trimethylammonium-propane, Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,2-Dioleoyl-3-trimethylammonium-propane, Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Tocris Bioscience™ DOTAP
Cationic agent for liposome and LNP formation
4
1,2-Dioleoyl-3-trimethylammonium-propane, Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.