Få mere at vide
Tocris Bioscience™ DOTAP
Beskrivelse
DOTAP is a cationic agent used in liposome formation. Facilitates the transfection of DNA, RNA, and other molecules with negative charges into eukaryotic cells. Can be used to generate selective organ-targeting (SORT) lipid nanoparticles. For more information on how to prepare MC3 lipid nanoparticles for RNA delivery, please see our protocol .
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N,N,N-Trimethyl-2,3-bis(oleoyloxy)propan-1-aminium chloride |
| CAS | 132172-61-3 |
| Mængde | 50 mg |
| Mål | Lipid Nanoparticles |
| Molekylær formel | C42H80ClNO4 |
| Renhed | 95% |