CAS RN 13214-66-9
IUPAC Navn:
4-phenylbutan-1-amine
Synonymer:
4-phenylbutylamine
Molekylvægt (g/mol):
149.24
Molekylformel:
C10H15N
InChi Key:
AGNFWIZBEATIAK-UHFFFAOYSA-N
SMILES:
NCCCCC1=CC=CC=C1
1
–
3
af
3
Resultater
1
4-Phenylbutylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Phenylbutylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Phenylbutylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.