CAS RN 131918-74-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(11Beta,16Alpha)-21-(Acetyloxy)-9-fluoro-11,16,17-trihydroxy-pregna-1,4,14-triene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(11Beta,16Alpha)-21-(Acetyloxy)-9-fluoro-11,16,17-trihydroxy-pregna-1,4,14-triene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.