CAS RN 131833-90-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Bis[(S)-4-Isopropyl-4,5-dihydrooxazol-2-yl]methane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Bis[(S)-4-Isopropyl-4,5-dihydrooxazol-2-yl]methane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Bis[(S)-4-Isopropyl-4,5-dihydrooxazol-2-yl]methane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.